MilliporeSigma
  • Tramiprosate, a drug of potential interest for the treatment of Alzheimer's disease, promotes an abnormal aggregation of tau.
Tramiprosate, a drug of potential interest for the treatment of Alzheimer's disease, promotes an abnormal aggregation of tau.
Molecular neurodegeneration (2007-09-08)
Ismael Santa-Maria, Félix Hernández, Joaquín Del Rio, Francisco J Moreno, Jesús Avila
ABSTRACT
Alzheimer's disease (AD) is characterized by the presence of two histopathological hallmarks; the senile plaques, or extracellular deposits mainly composed of amyloid-beta peptide (Abeta), and the neurofibrillary tangles, or intraneuronal inclusions composed of hyperphosphorylated tau protein.Since Abeta aggregates are found in the pathological cases, several strategies are under way to develop drugs that interact with Abeta to reduce its assembly. One of them is 3-amino-1-propane sulfonic acid (Tramiprosate, 3-APS, Alzhemedtrade mark), that was developed as a sulfated glycosaminoglycan mimetic, that could interact with Abeta peptide, preventing its aggregation.However, little is known about the action of 3-APS on tau protein aggregation. In this work, we have tested the action of 3-APS on cell viability, microtubule network, actin organization and tau aggregation. Our results indicate that 3-APS favours tau aggregation, in tau transfected non-neuronal cells, and in neuronal cells. We also found that 3-APS does not affect the binding of tau to microtubules but may prevent the formation of tau-actin aggregates. We like to emphasize the importance of testing on both types of pathology (amyloid and tau) the potential drugs to be used for AD treatment.
Sigma-Aldrich
Thioflavine S, practical grade
Sigma-Aldrich
DAPI, Dihydrochloride, Cell-permeable DNA-binding dye.
Millipore
ReBlot Plus Kit