MilliporeSigma
  • Gamma-secretase inhibition combined with platinum compounds enhances cell death in a large subset of colorectal cancer cells.
Gamma-secretase inhibition combined with platinum compounds enhances cell death in a large subset of colorectal cancer cells.
Cell communication and signaling : CCS (2008-10-28)
Tamara Aleksic, Stephan M Feller
ABSTRACT
Notch signalling is essential for the development and maintenance of the colonic epithelium. Its inhibition induces a differentiation phenotype in vivo and reduces adenomas in APCmin mice. Whether Notch signals are also required in colorectal cancer (CRC) has remained elusive. Therefore, 64 CRC cell lines were analysed for the occurrence of proteolytically processed, active Notch. 63 CRC lines contained a fragment with approximately the size of the Notch1 intracellular domain (NICD), which is required for signalling. Subsequent analyses with an antibody that specifically recognises the free Val1744 residue generated by gamma-secretase-mediated cleavage of Notch1 showed that a subset of CRC cells lacks this specific Val1744-NICD. Surprisingly, inhibition of Val1744-NICD signalling with different gamma-secretase inhibitors (GSI) did not lead to substantial effects on CRC cell line growth or survival. However, transient activation of Erk upon GSI treatment was detected. Since cisplatin relies on Erk activation for bioactivity in some cells, platinum compounds were tested together with GSI and enhanced cell killing in a subset of Val1744-NICD-positive CRC cell lines was detected. Erk inhibition ablated this combination effect. We conclude that gamma-secretase inhibition results in activation of the MAP kinases Erk1/2 and, when used in conjunction, enhances cell death induced by platinum compounds in a large subset of colorectal cancer cell lines.Furthermore the activation of Erk appears to be of particular importance in mediating the enhanced effect seen, as its inhibition abrogates the observed phenomenon. These findings do not only highlight the importance of signalling pathway crosstalk but they may also suggest a new avenue of combination therapy for some colorectal cancers.
Sigma-Aldrich
Anti-Actin antibody, Mouse monoclonal, clone AC-40, purified from hybridoma cell culture
Sigma-Aldrich
γ-Secretase Inhibitor IX, Gamma-Secretase Inhibitor IX - CAS 208255-80-5, is a cell-permeable inhibitor of γ-secretase (Aβtotal IC₅₀ = 115 nM, Aβ42 IC₅₀ = 200 nM).
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in glass vials
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Anti-Notch1 antibody, Mouse monoclonal, clone mN1A, purified from hybridoma cell culture
Sigma-Aldrich
Cisplatin, Cisplatin - CAS 15663-27-1, is a platinum coordination complex with potent anti-neoplastic activity. Induces apoptosis in cancer cells, possibly via caspase-3 activation.
Sigma-Aldrich
γ-Secretase Inhibitor XX, This γ-secretase inhibitor, CAS 209984-56-5, is a cell-permeable dibenzazepine compound that lowers both brain and plasma Aβ40 levels by ~72% in Tg2576 mutant APP transgenic mouse model.