MilliporeSigma
  • Pyruvate carboxylation enables growth of SDH-deficient cells by supporting aspartate biosynthesis.
Pyruvate carboxylation enables growth of SDH-deficient cells by supporting aspartate biosynthesis.
Nature cell biology (2015-08-25)
Simone Cardaci, Liang Zheng, Gillian MacKay, Niels J F van den Broek, Elaine D MacKenzie, Colin Nixon, David Stevenson, Sergey Tumanov, Vinay Bulusu, Jurre J Kamphorst, Alexei Vazquez, Stewart Fleming, Francesca Schiavi, Gabriela Kalna, Karen Blyth, Douglas Strathdee, Eyal Gottlieb
ABSTRACT
Succinate dehydrogenase (SDH) is a heterotetrameric nuclear-encoded complex responsible for the oxidation of succinate to fumarate in the tricarboxylic acid cycle. Loss-of-function mutations in any of the SDH genes are associated with cancer formation. However, the impact of SDH loss on cell metabolism and the mechanisms enabling growth of SDH-defective cells are largely unknown. Here, we generated Sdhb-ablated kidney mouse cells and used comparative metabolomics and stable-isotope-labelling approaches to identify nutritional requirements and metabolic adaptations to SDH loss. We found that lack of SDH activity commits cells to consume extracellular pyruvate, which sustains Warburg-like bioenergetic features. We further demonstrated that pyruvate carboxylation diverts glucose-derived carbons into aspartate biosynthesis, thus sustaining cell growth. By identifying pyruvate carboxylase as essential for the proliferation and tumorigenic capacity of SDH-deficient cells, this study revealed a metabolic vulnerability for potential future treatment of SDH-associated malignancies.
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-PC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution