MilliporeSigma
  • Synthesis, characterization, and photovoltaic properties of soluble TiOPc derivatives.
Synthesis, characterization, and photovoltaic properties of soluble TiOPc derivatives.
International journal of molecular sciences (2009-03-31)
Young-Keun Kim, Hyo-Jin Kang, Young-Wook Jang, Su-Bin Lee, Seung-Min Lee, Ki-Suck Jung, Jin-Kook Lee, Mi-Ra Kim
ABSTRACT
We have synthesized soluble TiOPc derivatives containing alkoxy groups for use as additives in dye-sensitized solar cells (DSSCs). The DSSC devices containing these TiOPc derivatives exhibited short-circuit current densities of 8.49~10.04 mA/cm(2) and power conversion efficiencies of 2.73~3.62 % under AM 1.5 illumination and 100 mW/cm(2) irradiation.