MilliporeSigma
  • Microclusters of inhibitory killer immunoglobulin-like receptor signaling at natural killer cell immunological synapses.
Microclusters of inhibitory killer immunoglobulin-like receptor signaling at natural killer cell immunological synapses.
The Journal of cell biology (2006-06-28)
Bebhinn Treanor, Peter M P Lanigan, Sunil Kumar, Chris Dunsby, Ian Munro, Egidijus Auksorius, Fiona J Culley, Marco A Purbhoo, David Phillips, Mark A A Neil, Deborah N Burshtyn, Paul M W French, Daniel M Davis
ABSTRACT
We report the supramolecular organization of killer Ig-like receptor (KIR) phosphorylation using a technique applicable to imaging phosphorylation of any green fluorescent protein-tagged receptor at an intercellular contact or immune synapse. Specifically, we use fluorescence lifetime imaging (FLIM) to report Förster resonance energy transfer (FRET) between GFP-tagged KIR2DL1 and a Cy3-tagged generic anti-phosphotyrosine monoclonal antibody. Visualization of KIR phosphorylation in natural killer (NK) cells contacting target cells expressing cognate major histocompatibility complex class I proteins revealed that inhibitory signaling is spatially restricted to the immune synapse. This explains how NK cells respond appropriately when simultaneously surveying susceptible and resistant target cells. More surprising, phosphorylated KIR was confined to microclusters within the aggregate of KIR, contrary to an expected homogeneous distribution of KIR signaling across the immune synapse. Also, yellow fluorescent protein-tagged Lck, a kinase important for KIR phosphorylation, accumulated in a multifocal distribution at inhibitory synapses. Spatial confinement of receptor phosphorylation within the immune synapse may be critical to how activating and inhibitory signals are integrated in NK cells.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
7-Cyclopentyl-5-(4-phenoxyphenyl)-7H-pyrrolo[2,3‑d]pyrimidin-4-ylamine, ≥98% (HPLC)