MilliporeSigma
  • The Three Catecholics Benserazide, Catechol and Pyrogallol are GPR35 Agonists.
The Three Catecholics Benserazide, Catechol and Pyrogallol are GPR35 Agonists.
Pharmaceuticals (Basel, Switzerland) (2013-11-28)
Huayun Deng, Ye Fang
ABSTRACT
Nearly 1% of all clinically used drugs are catecholics, a family of catechol-containing compounds. Using label-free dynamic mass redistribution and Tango β-arrestin translocation assays, we show that several catecholics, including benserazide, catechol, 3-methoxycatechol, pyrogallol, (+)-taxifolin and fenoldopam, display agonistic activity against GPR35.
Sigma-Aldrich
3-Methoxycatechol, 99%