MilliporeSigma
  • Palmitic acid and ergosta-7,22-dien-3-ol contribute to the apoptotic effect and cell cycle arrest of an extract from Marthasterias glacialis L. in neuroblastoma cells.
Palmitic acid and ergosta-7,22-dien-3-ol contribute to the apoptotic effect and cell cycle arrest of an extract from Marthasterias glacialis L. in neuroblastoma cells.
Marine drugs (2013-12-26)
David M Pereira, Georgina Correia-da-Silva, Patrícia Valentão, Natércia Teixeira, Paula B Andrade
ABSTRACT
We describe the effect of a chemically characterized lipophilic extract obtained from Marthasterias glacialis L. against human breast cancer (MCF-7) and human neuroblastoma (SH-SY5Y) cell lines. Evaluation of DNA synthesis revealed that both cell lines were markedly affected in a concentration-dependent way, the SH-SY5Y cell line being more susceptible. Cell cycle arrest was observed, an effect induced by the sterol, ergosta-7,22-dien-3-ol, present in the extract. Morphological evaluation of treated cells showed the advent of lipid droplets and chromatin condensation compatible with apoptosis, which was confirmed by the evaluation of caspase-3 and -9 activities. Palmitic acid was the main compound responsible for this apoptotic effect by a ceramide-independent mechanism that involved endoplasmic reticulum (ER)-stress with upregulation of CCAAT/-enhancer-binding protein homologous protein (CHOP).
Sigma-Aldrich
L-Cycloserine