MilliporeSigma
  • Discovery of a structural-element specific G-quadruplex "light-up" probe.
Discovery of a structural-element specific G-quadruplex "light-up" probe.
Scientific reports (2014-01-21)
Liyun Zhang, Jun Cheng Er, Krishna Kanta Ghosh, Wan Jun Chung, Jaeduk Yoo, Wang Xu, Wei Zhao, Anh Tuân Phan, Young-Tae Chang
ABSTRACT
The development of a fluorescent probe capable of detecting and distinguishing the wide diversity of G-quadruplex structures is particularly challenging. Herein, we report a novel BODIPY-based fluorescent sensor (GQR) that shows unprecedented selectivity to parallel-stranded G-quadruplexes with exposed ends and four medium grooves. Mechanistic studies suggest that GQR associates with G-quadruplex grooves close to the end of the tetrad core, which may explain the dye's specificity to only a subset of parallel structures. This specific recognition favours the disaggregation of GQR in aqueous solutions thereby recovering the inherent fluorescence of the dye. Due to its unique features, GQR represents a valuable tool for basic biological research and the rapid discovery of novel, specific ligands that target similar structural features of G-quadruplexes.
Sigma-Aldrich
Deoxyribonucleic acid−cellulose double-stranded from calf thymus DNA, lyophilized powder
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from calf thymus, lyophilized powder