MilliporeSigma
  • Discovery of 3-alkoxyamino-5-(pyridin-2-ylamino)pyrazine-2-carbonitriles as selective, orally bioavailable CHK1 inhibitors.
Discovery of 3-alkoxyamino-5-(pyridin-2-ylamino)pyrazine-2-carbonitriles as selective, orally bioavailable CHK1 inhibitors.
Journal of medicinal chemistry (2012-10-23)
Michael Lainchbury, Thomas P Matthews, Tatiana McHardy, Kathy J Boxall, Michael I Walton, Paul D Eve, Angela Hayes, Melanie R Valenti, Alexis K de Haven Brandon, Gary Box, G Wynne Aherne, John C Reader, Florence I Raynaud, Suzanne A Eccles, Michelle D Garrett, Ian Collins
ABSTRACT
Inhibitors of checkpoint kinase 1 (CHK1) are of current interest as potential antitumor agents, but the most advanced inhibitor series reported to date are not orally bioavailable. A novel series of potent and orally bioavailable 3-alkoxyamino-5-(pyridin-2-ylamino)pyrazine-2-carbonitrile CHK1 inhibitors was generated by hybridization of two lead scaffolds derived from fragment-based drug design and optimized for CHK1 potency and high selectivity using a cell-based assay cascade. Efficient in vivo pharmacokinetic assessment was used to identify compounds with prolonged exposure following oral dosing. The optimized compound (CCT244747) was a potent and highly selective CHK1 inhibitor, which modulated the DNA damage response pathway in human tumor xenografts and showed antitumor activity in combination with genotoxic chemotherapies and as a single agent.
Sigma-Aldrich
1-Methylpyrazole-4-boronic acid pinacol ester, 95%