MilliporeSigma
  • Surface-immobilization of chromatographically purified bacteriophages for the optimized capture of bacteria.
Surface-immobilization of chromatographically purified bacteriophages for the optimized capture of bacteria.
Bacteriophage (2012-06-06)
Ravendra Naidoo, Amit Singh, Sunil K Arya, Bernadette Beadle, Nick Glass, Jamshid Tanha, Christine M Szymanski, Stephane Evoy
ABSTRACT
Bacteriophages offer interesting alternatives to antibodies for the specific capture and detection of pathogenic bacteria onto biosensing surfaces. Procedures for the optimal chemical immobilization of lytic bacteriophages onto surfaces are presented. More specifically, the removal of lysate contaminants from bacteriophage suspensions by size exclusion chromatography significantly increases the resultant planar surface density of immobilized bacteriophages. E. coli T4 and Salmonella enterica serovar Typhimurium P22 phage systems seem to undergo highly heterogeneous adsorption to the surface, possibly explaining the observed phage clustering at higher surface densities. The T4 phage and its E. coli host were initially employed as a model system where we discovered an optimal planar surface density of phages for best bacterial capture: 18.9 ± 0.8 phages/μm(2) capturing 18.0 ± 0.3 bacteria/100 μm(2). Phage surface clustering ultimately limits the T4 phage-immobilized surface's ability to specifically capture its host bacteria. Nevertheless, this is to our knowledge the largest surface capture density of E. coli reported using intact T4 bacteriophages. Two additional purified bacteriophage systems (P22 and Campylobacter jejuni phage NCTC 12673) were then similarly studied for their ability to capture their corresponding host bacteria (Salmonella enterica serovar Typhimurium and Campylobacter jejuni respectively) on a surface.
Sigma-Aldrich
Cysteamine, ~95%