MilliporeSigma
Flexible antibodies with nonprotein hinges.
Proceedings of the Japan Academy. Series B, Physical and biological sciences (2011-11-15)
Daniel J Capon, Naoki Kaneko, Takayuki Yoshimori, Takashi Shimada, Florian M Wurm, Peter K Hwang, Xiaohe Tong, Staci A Adams, Graham Simmons, Taka-Aki Sato, Koichi Tanaka
ABSTRACT
There is a significant need for antibodies that can bind targets with greater affinity. Here we describe a novel strategy employing chemical semisynthesis to produce symmetroadhesins: antibody-like molecules having nonprotein hinge regions that are more flexible and extendible and are capable of two-handed binding. Native chemical ligation was carried out under mild, non-denaturing conditions to join a ligand binding domain (Aβ peptide) to an IgG1 Fc dimer via discrete oxyethylene oligomers of various lengths. Two-handed Aβ-Fc fusion proteins were obtained in quantitative yield and shown by surface plasmon resonance to bind an anti-Aβ antibody with a K(D) at least two orders of magnitude greater than the cognate Aβ peptide. MALDI-TOF MS analysis confirmed the protein/nonprotein/protein structure of the two-handed molecules, demonstrating its power to characterize complex protein-nonprotein hybrids by virtue of desorption/ionization mediated by peptide sequences contained therein. We anticipate many applications for symmetroadhesins that combine the target specificity of antibodies with the novel physical, chemical and biological properties of nonprotein hinges.
Sigma-Aldrich
4-Mercaptophenylacetic acid, 97%