MilliporeSigma
  • Kinases Mst1 and Mst2 positively regulate phagocytic induction of reactive oxygen species and bactericidal activity.
Kinases Mst1 and Mst2 positively regulate phagocytic induction of reactive oxygen species and bactericidal activity.
Nature immunology (2015-09-29)
Jing Geng, Xiufeng Sun, Ping Wang, Shihao Zhang, Xiaozhen Wang, Hongtan Wu, Lixin Hong, Changchuan Xie, Xun Li, Hao Zhao, Qingxu Liu, Mingting Jiang, Qinghua Chen, Jinjia Zhang, Yang Li, Siyang Song, Hong-Rui Wang, Rongbin Zhou, Randy L Johnson, Kun-Yi Chien, Sheng-Cai Lin, Jiahuai Han, Joseph Avruch, Lanfen Chen, Dawang Zhou
ABSTRACT
Mitochondria need to be juxtaposed to phagosomes for the synergistic production of ample reactive oxygen species (ROS) in phagocytes to kill pathogens. However, how phagosomes transmit signals to recruit mitochondria has remained unclear. Here we found that the kinases Mst1 and Mst2 functioned to control ROS production by regulating mitochondrial trafficking and mitochondrion-phagosome juxtaposition. Mst1 and Mst2 activated the GTPase Rac to promote Toll-like receptor (TLR)-triggered assembly of the TRAF6-ECSIT complex that is required for the recruitment of mitochondria to phagosomes. Inactive forms of Rac, including the human Rac2(D57N) mutant, disrupted the TRAF6-ECSIT complex by sequestering TRAF6 and substantially diminished ROS production and enhanced susceptibility to bacterial infection. Our findings demonstrate that the TLR-Mst1-Mst2-Rac signaling axis is critical for effective phagosome-mitochondrion function and bactericidal activity.
Sigma-Aldrich
UEV1A active human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)