MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • RNAi-mediated abrogation of trehalase expression does not affect trehalase activity in sugarcane.
RNAi-mediated abrogation of trehalase expression does not affect trehalase activity in sugarcane.
SpringerPlus (2013-03-30)
Brian P O'Neill, Matthew P Purnell, Lars K Nielsen, Stevens M Brumbley
ABSTRACT
To engineer trehalose metabolism in sugarcane (Saccharum spp. hybrids) two transgenes were introduced to the genome: trehalose-6-phosphate synthase- phosphatase (TPSP), to increase trehalose biosynthesis and an RNAi transgene specific for trehalase, to abrogate trehalose catabolism. In RNAi-expressing lines trehalase expression was abrogated in many plants however no decrease in trehalase activity was observed. In TPSP lines trehalase activity was significantly higher. No events of co-integration of TPSP and RNAi transgenes were observed. We suggest trehalase activity is essential to mitigate embryonic lethal effects of trehalose metabolism and discuss the implications for engineering trehalose metabolism.