MilliporeSigma
Structural antitumoral activity relationships of synthetic chalcones.
International journal of molecular sciences (2009-04-01)
Cesar Echeverria, Juan Francisco Santibañez, Oscar Donoso-Tauda, Carlos A Escobar, Rodrigo Ramirez-Tagle
ABSTRACT
Relationships between the structural characteristic of synthetic chalcones and their antitumoral activity were studied. Treatment of HepG2 cells for 24 h with synthetic 2'-hydroxychalcones resulted in apoptosis induction and dose-dependent inhibition of cell proliferation. The calculated reactivity indexes and the adiabatic electron affinities using the DFT method including solvent effects, suggest a structure-activity relationship between the Chalcones structure and the apoptosis in HepG2 cells. The absence of methoxy substituents in the B ring of synthetic 2'-hydroxychalcones, showed the mayor structure-activity pattern along the series.