MilliporeSigma
  • Degradation of Iodinated Contrast Media in Aquatic Environment by Means of UV, UV/TiO
Degradation of Iodinated Contrast Media in Aquatic Environment by Means of UV, UV/TiO
Water, air, and soil pollution (2015-05-12)
Ewa Borowska, Ewa Felis, Sebastian Żabczyński
ABSTRACT
Iodinated contrast media (ICM), which are used for radiological visualization of human tissue and cardiovascular system, are poorly biodegradable; hence, new methods of their removal are sought. In this study, the effectiveness of selected X-ray ICM removal by means of UV and UV/TiO