MilliporeSigma
  • Loss of miR-200b promotes invasion via activating the Kindlin-2/integrin β1/AKT pathway in esophageal squamous cell carcinoma: An E-cadherin-independent mechanism.
Loss of miR-200b promotes invasion via activating the Kindlin-2/integrin β1/AKT pathway in esophageal squamous cell carcinoma: An E-cadherin-independent mechanism.
Oncotarget (2015-09-04)
Hai-Feng Zhang, Abdulraheem Alshareef, Chengsheng Wu, Shang Li, Ji-Wei Jiao, Hui-Hui Cao, Raymond Lai, Li-Yan Xu, En-Min Li
ABSTRACT
Our previous studies have shown that loss of miR-200b enhances the invasiveness of esophageal squamous cell carcinoma (ESCC) cells. However, whether the miR-200-ZEB1/2-E-cadherin regulatory cascade, a master regulator of epithelial-to-mesenchymal transition (EMT), is involved in the regulation of ESCC invasion remains elusive. Here, we show that miR-200b represses ESCC cell invasion in vivo without altering the expression of E-cadherin and vimentin, two surrogate markers of EMT. However, an inverse correlation was observed between the expression levels of miR-200b and ZEB1/2 in both ESCC cell lines (n = 7, P < 0.05) and ESCC tumor samples (n = 88, P < 0.05). Methylation of E-cadherin gene was found to block the regulation of E-cadherin by the miR-200b-ZEB1/2 axis, indicating that an E-cadherin-independent mechanism can mediate the biological function of miR-200b in ESCC. We revealed that miR-200b suppresses the integrin β1-AKT pathway via targeting Kindlin-2 to mitigate ESCC cell invasiveness. In two independent cohorts of ESCC samples (n = 20 and n = 53, respectively), Kindlin-2 expression positively correlated with the activation status of both the integrin signaling pathway and the PI3K-AKT signaling pathway (both P < 0.01). These data highlight that suppression of the Kindlin-2-integrin β1-AKT regulatory axis is an alternative mechanism underlying the tumor suppressor function of miR-200b in ESCC.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fermt2
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%