MilliporeSigma
  • Effects of Dietary Supplementation of Barodon, an Anionic Alkali Mineral Complex, on Growth Performance, Feed Utilization, Innate Immunity, Goblet Cell and Digestibility in Olive Flounder (Paralichthys olivaceus).
Effects of Dietary Supplementation of Barodon, an Anionic Alkali Mineral Complex, on Growth Performance, Feed Utilization, Innate Immunity, Goblet Cell and Digestibility in Olive Flounder (Paralichthys olivaceus).
Asian-Australasian journal of animal sciences (2014-07-23)
Chang-Hoon Shin, Ji-Hoon Cha, Samad Rahimnejad, Joon-Bum Jeong, Byung-Woo Yoo, Bo-Kyeun Lee, Hyung-Jin Ahn, Soo-Il Choi, Yun-Jeong Choi, Yong-Ho Park, Jeong-Dae Kim, Kyeong-Jun Lee
ABSTRACT
A 15-wk feeding trial was conducted to examine the supplemental effects of Barodon on growth performance, gastrointestinal histology, feed digestibility and innate immunity in olive founder. A basal commercial diet was used as a control and two other diets were prepared by spraying 0.1% or 0.2% of Barodon. Triplicate groups of fish (BW, 145 g) were fed one of the test diets to apparent satiation twice daily. At the end of the feeding trial, fish growth performance was not significantly affected by dietary treatments; however, feed utilization was significantly improved (linear and quadratic, p<0.05) by Barodon supplementation. Significantly higher (p<0.05) survival rates were obtained in fish fed Barodon containing diets. Hepatosomatic index increased significantly in Barodon treated groups. Also, the use of Barodon resulted in significant increase (linear and quadratic, p<0.05) of intestine length and number of goblet cells. Significantly higher (Quadratic, p<0.05) apparent digestibility coefficient of DM was obtained by supplementation of Barodon. Lysozyme and myeloperoxidase activities increased quadratically and linearly, respectively, in Barodon treated fish. Also, significantly higher (linear and quadratic, p<0.05) superoxide dismutase activity was found in Barodon fed fish. The findings in this study show that inclusion of Barodon in diets for olive flounder improves feed utilization and digestibility, and positively affects digestive tract histology and innate immunity.
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests
Sigma-Aldrich
BrdU (5-Bromo-2′-deoxyuridine), Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine, BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.