MilliporeSigma
  • Functional comparison of single-chain and two-chain anti-CD3-based bispecific antibodies in gene immunotherapy applications.
Functional comparison of single-chain and two-chain anti-CD3-based bispecific antibodies in gene immunotherapy applications.
Oncoimmunology (2014-07-25)
Marta Compte, Ana Alvarez-Cienfuegos, Natalia Nuñez-Prado, Noelia Sainz-Pastor, Ana Blanco-Toribio, Nuria Pescador, Laura Sanz, Luis Alvarez-Vallina
ABSTRACT
Gene therapy to achieve in vivo secretion of recombinant anti-CD3 x anti-tumor bispecific antibodies in cancer patients is being explored as a strategy to counterbalance rapid renal elimination, thereby sustaining levels of bispecific antibodies in the therapeutic range. Here, we performed a comparative analysis between single- and two-chain configurations for anti-CD3 x anti-CEA (carcinoembryonic antigen) bispecific antibodies secreted by genetically-modified human cells. We demonstrate that tandem single-chain variable fragment (scFv) antibodies and two-chain diabodies are expressed as soluble secreted proteins with similar yields. However, we found significant differences in their biological functionality (i.e., antigen binding) and in their ability to induce non-specific T cell activation. Whereas single-chain tandem scFvs induced human T cell activation and proliferation in an antigen-independent manner, secreted two-chain diabodies exerted almost no proliferative stimulus when human T cells were cultured alone or in co-cultures with CEA negative cells. Thus, our data suggest that two-chain diabodies are preferable to single-chain tandem scFvs for immunotherapeutic strategies comprising in vivo secretion of bispecific antibodies aiming to recruit and activate anticancer specific lymphocytic effector T cells.
Sigma-Aldrich
Monoclonal Anti-Carcinoembryonic Antigen antibody produced in mouse, clone C6G9, ascites fluid