MilliporeSigma
  • The role of paracrine and autocrine signaling in the early phase of adipogenic differentiation of adipose-derived stem cells.
The role of paracrine and autocrine signaling in the early phase of adipogenic differentiation of adipose-derived stem cells.
PloS one (2013-06-01)
Mette Hemmingsen, Søren Vedel, Peder Skafte-Pedersen, David Sabourin, Philippe Collas, Henrik Bruus, Martin Dufva
ABSTRACT
High cell density is known to enhance adipogenic differentiation of mesenchymal stem cells, suggesting secretion of signaling factors or cell-contact-mediated signaling. By employing microfluidic biochip technology, we have been able to separate these two processes and study the secretion pathways. Adipogenic differentiation of human adipose-derived stem cells (ASCs) cultured in a microfluidic system was investigated under perfusion conditions with an adipogenic medium or an adipogenic medium supplemented with supernatant from differentiating ASCs (conditioned medium). Conditioned medium increased adipogenic differentiation compared to adipogenic medium with respect to accumulation of lipid-filled vacuoles and gene expression of key adipogenic markers (C/EBPα, C/EBPβ, C/EBPδ, PPARγ, LPL and adiponectin). The positive effects of conditioned medium were observed early in the differentiation process. Using different cell densities and microfluidic perfusion cell cultures to suppress the effects of cell-released factors, we have demonstrated the significant role played by auto- or paracrine signaling in adipocyte differentiation. The cell-released factor(s) were shown to act in the recruitment phase of the differentiation process.
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Newborn Calf Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture