MilliporeSigma
  • A gp41 MPER-specific llama VHH requires a hydrophobic CDR3 for neutralization but not for antigen recognition.
A gp41 MPER-specific llama VHH requires a hydrophobic CDR3 for neutralization but not for antigen recognition.
PLoS pathogens (2013-03-19)
David Lutje Hulsik, Ying-ying Liu, Nika M Strokappe, Simone Battella, Mohamed El Khattabi, Laura E McCoy, Charles Sabin, Andreas Hinz, Miriam Hock, Pauline Macheboeuf, Alexandre M J J Bonvin, Johannes P M Langedijk, David Davis, Anna Forsman Quigley, Marlén M I Aasa-Chapman, Michael S Seaman, Alejandra Ramos, Pascal Poignard, Adrien Favier, Jean-Pierre Simorre, Robin A Weiss, C Theo Verrips, Winfried Weissenhorn, Lucy Rutten
ABSTRACT
The membrane proximal external region (MPER) of the HIV-1 glycoprotein gp41 is targeted by the broadly neutralizing antibodies 2F5 and 4E10. To date, no immunization regimen in animals or humans has produced HIV-1 neutralizing MPER-specific antibodies. We immunized llamas with gp41-MPER proteoliposomes and selected a MPER-specific single chain antibody (VHH), 2H10, whose epitope overlaps with that of mAb 2F5. Bi-2H10, a bivalent form of 2H10, which displayed an approximately 20-fold increased affinity compared to the monovalent 2H10, neutralized various sensitive and resistant HIV-1 strains, as well as SHIV strains in TZM-bl cells. X-ray and NMR analyses combined with mutagenesis and modeling revealed that 2H10 recognizes its gp41 epitope in a helical conformation. Notably, tryptophan 100 at the tip of the long CDR3 is not required for gp41 interaction but essential for neutralization. Thus bi-2H10 is an anti-MPER antibody generated by immunization that requires hydrophobic CDR3 determinants in addition to epitope recognition for neutralization similar to the mode of neutralization employed by mAbs 2F5 and 4E10.
Sigma-Aldrich
1,2-Diacyl-sn-glycero-3-phospho-L-serine, ≥97% (TLC), from bovine brain, amorphous powder
Sigma-Aldrich
Cardiolipin sodium salt from bovine heart, ≥97% (TLC), lyophilized powder