MilliporeSigma
  • K+ channel regulator KCR1 suppresses heart rhythm by modulating the pacemaker current If.
K+ channel regulator KCR1 suppresses heart rhythm by modulating the pacemaker current If.
PloS one (2008-01-31)
Guido Michels, Fikret Er, Ismail F Khan, Jeannette Endres-Becker, Mathias C Brandt, Natig Gassanov, David C Johns, Uta C Hoppe
ABSTRACT
Hyperpolarization-activated, cyclic nucleotide sensitive (HCN) channels underlie the pacemaker current I(f), which plays an essential role in spontaneous cardiac activity. HCN channel subunits (HCN1-4) are believed to be modulated by additional regulatory proteins, which still have to be identified. Using biochemistry, molecularbiology and electrophysiology methods we demonstrate a protein-protein interaction between HCN2 and the K(+) channel regulator protein 1, named KCR1. In coimmunoprecipitation experiments we show that KCR1 and HCN2 proteins are able to associate. Heterologously expressed HCN2 whole-cell current density was significantly decreased by KCR1. KCR1 profoundly suppressed I(HCN2) single-channel activity, indicating a functional interaction between KCR1 and the HCN2 channel subunit. Endogenous KCR1 expression could be detected in adult and neonatal rat ventriculocytes. Adenoviral-mediated overexpression of KCR1 in rat cardiomyocytes (i) reduced I(f) whole-cell currents, (ii) suppressed most single-channel gating parameters, (iii) altered the activation kinetics, (iv) suppressed spontaneous action potential activity, and (v) the beating rate. More importantly, siRNA-based knock-down of endogenous KCR1 increased the native I(f) current size and single-channel activity and accelerated spontaneous beating rate, supporting an inhibitory action of endogenous KCR1 on native I(f). Our observations demonstrate for the first time that KCR1 modulates I(HCN2)/I(f) channel gating and indicate that KCR1 serves as a regulator of cardiac automaticity.
Sigma-Aldrich
8-Bromoadenosine 5′-monophosphate, ≥98%