MilliporeSigma
  • The non-JAZ TIFY protein TIFY8 from Arabidopsis thaliana is a transcriptional repressor.
The non-JAZ TIFY protein TIFY8 from Arabidopsis thaliana is a transcriptional repressor.
PloS one (2014-01-15)
Amparo Cuéllar Pérez, Astrid Nagels Durand, Robin Vanden Bossche, Rebecca De Clercq, Geert Persiau, Saskia C M Van Wees, Corné M J Pieterse, Kris Gevaert, Geert De Jaeger, Alain Goossens, Laurens Pauwels
ABSTRACT
Jasmonate (JA) signalling is mediated by the JASMONATE-ZIM DOMAIN (JAZ) repressor proteins, which are degraded upon JA perception to release downstream responses. The ZIM protein domain is characteristic of the larger TIFY protein family. It is currently unknown if the atypical member TIFY8 is involved in JA signalling. Here we show that the TIFY8 ZIM domain is functional and mediated interaction with PEAPOD proteins and NINJA. TIFY8 interacted with TOPLESS through NINJA and accordingly acted as a transcriptional repressor. TIFY8 expression was inversely correlated with JAZ expression during development and after infection with Pseudomonas syringae. Nevertheless, transgenic lines with altered TIFY8 expression did not show changes in JA sensitivity. Despite the functional ZIM domain, no interaction with JAZ proteins could be found. In contrast, TIFY8 was found in protein complexes involved in regulation of dephosphorylation, deubiquitination and O-linked N-acetylglucosamine modification suggesting an important role in nuclear signal transduction.
Sigma-Aldrich
Peroxidase Anti-Peroxidase Soluble Complex antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution