MilliporeSigma
  • Evaluation of an Ultrafiltration-Based Procedure for Simultaneous Recovery of Diverse Microbes in Source Waters.
Evaluation of an Ultrafiltration-Based Procedure for Simultaneous Recovery of Diverse Microbes in Source Waters.
Water (2015-11-05)
Amy M Kahler, Trisha B Johnson, Donghyun Hahn, Jothikumar Narayanan, Gordana Derado, Vincent R Hill
ABSTRACT
In this study, hollow-fiber ultrafiltration (UF) was assessed for recovery of
Sigma-Aldrich
Sodium hexametaphosphate, crystalline, +200 mesh, 96%