MilliporeSigma
  • Selective urokinase-type plasminogen activator (uPA) inhibitors. Part 3: 1-isoquinolinylguanidines.
Selective urokinase-type plasminogen activator (uPA) inhibitors. Part 3: 1-isoquinolinylguanidines.
Bioorganic & medicinal chemistry letters (2004-05-20)
Christopher G Barber, Roger P Dickinson, Paul V Fish
ABSTRACT
A series of 1-isoquinolinylguanidines are shown to be potent inhibitors of uPA with selectivity over tPA and plasmin. Potency is enhanced by the presence of a 4-halo and a 7-aryl substituent, particularly when substituted by a 3-carboxylic acid group. Compound 13j (UK-356,202) combines excellent potency and selectivity, and has been selected as a candidate for clinical evaluation.
Sigma-Aldrich
3-Carboxyphenylboronic acid, ≥95%