MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Double-chelation-assisted Rh-catalyzed intermolecular hydroacylation between salicylaldehydes and 1,4-penta- or 1,5-hexadienes.
Double-chelation-assisted Rh-catalyzed intermolecular hydroacylation between salicylaldehydes and 1,4-penta- or 1,5-hexadienes.
The Journal of organic chemistry (2004-02-14)
Masanori Imai, Masakazu Tanaka, Keitaro Tanaka, Yoichiro Yamamoto, Naoko Imai-Ogata, Masato Shimowatari, Shinji Nagumo, Norio Kawahara, Hiroshi Suemune
ABSTRACT
Intermolecular hydroacylation between salicylaldehydes 1, 26-40 and 1,4-penta- or 1,5-hexadienes 4-13 by Rh-catalyst proceeded under mild reaction conditions to give a mixture of iso- and normal-hydroacylated products 14-25, 41-55, and 57-60. In the hydroacylation reaction, chelation of both salicylaldehyde and diene to the Rh-complex plays a crucial role. The ratio of iso- and normal-hydroacylated products could be regulated by the addition of salicylic acid or amines. The effects of various Rh-complexes, solvents, and additives were examined, and the plausible mechanisms of the catalytic cycle were proposed on the basis of the deuterium-labeling salicylaldehyde experiments.
Sigma-Aldrich
1,4-Pentadien-3-ol, contains 0.4% hydroquinone as stabilizer, ≥96%