MilliporeSigma
  • The NALP3 inflammasome is involved in neurotoxic prion peptide-induced microglial activation.
The NALP3 inflammasome is involved in neurotoxic prion peptide-induced microglial activation.
Journal of neuroinflammation (2012-04-26)
Fushan Shi, Lifeng Yang, Mohammed Kouadir, Yang Yang, Jihong Wang, Xiangmei Zhou, Xiaomin Yin, Deming Zhao
ABSTRACT
Prion diseases are neurodegenerative disorders characterized by the accumulation of an abnormal disease-associated prion protein, PrPSc. In prion-infected brains, activated microglia are often present in the vicinity of PrPSc aggregates, and microglial activation is thought to play a key role in the pathogenesis of prion diseases. Although interleukin (IL)-1β release by prion-induced microglia has been widely reported, the mechanism by which primed microglia become activated and secrete IL-1β in prion diseases has not yet been elucidated. In this study, we investigated the role of the NACHT, LRR and PYD domains-containing protein (NALP)3 inflammasome in IL-1β release from lipopolysaccharide (LPS)-primed microglia after exposure to a synthetic neurotoxic prion fragment (PrP106-126). The inflammasome components NALP3 and apoptosis-associated speck-like protein (ASC) were knocked down by gene silencing. IL-1β production was assessed using ELISA. The mRNA expression of NALP3, ASC, and pro-inflammatory factors was measured by quantitative PCR. Western blot analysis was used to detect the protein level of NALP3, ASC, caspase-1 and nuclear factor-κB. We found that that PrP106-126-induced IL-1β release depends on NALP3 inflammasome activation, that inflammasome activation is required for the synthesis of pro-inflammatory and chemotactic factors by PrP106-126-activated microglia, that inhibition of NF-κB activation abrogated PrP106-126-induced NALP3 upregulation, and that potassium efflux and production of reactive oxygen species were implicated in PrP106-126-induced NALP3 inflammasome activation in microglia. We conclude that the NALP3 inflammasome is involved in neurotoxic prion peptide-induced microglial activation. To our knowledge, this is the first time that strong evidence for the involvement of NALP3 inflammasome in prion-associated inflammation has been found.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction