MilliporeSigma
  • Activation of IGF1R/p110β/AKT/mTOR confers resistance to α-specific PI3K inhibition.
Activation of IGF1R/p110β/AKT/mTOR confers resistance to α-specific PI3K inhibition.
Breast cancer research : BCR (2016-04-07)
Cedric Leroy, Pedro Ramos, Karen Cornille, Debora Bonenfant, Christine Fritsch, Hans Voshol, Mohamed Bentires-Alj
ABSTRACT
The PI3K pathway is hyperactivated in many cancers, including 70 % of breast cancers. Pan- and isoform-specific inhibitors of the PI3K pathway are currently being evaluated in clinical trials. However, the clinical responses to PI3K inhibitors when used as single agents are not as efficient as expected. In order to anticipate potential molecular mechanisms of resistance to the p110α isoform-selective inhibitor BYL719, we developed resistant breast cancer cell lines, assessed the concomitant changes in cellular signaling pathways using unbiased phosphotyrosine proteomics and characterized the mechanism of resistance using pharmacological inhibitors. We found an increase in IGF1R, IRS1/IRS2 and p85 phosphorylation in the resistant lines. Co-immunoprecipitation experiments identified an IGF1R/IRS/p85/p110β complex that causes the activation of AKT/mTOR/S6K and stifles the effects of BYL719. Pharmacological inhibition of members of this complex reduced mTOR/S6K activation and restored sensitivity to BYL719. Our study demonstrates that the IGF1R/p110β/AKT/mTOR axis confers resistance to BYL719 in PIK3CA mutant breast cancers. This provides a rationale for the combined targeting of p110α with IGF1R or p110β in patients with breast tumors harboring PIK3CA mutations.
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
GSK2334470, ≥98% (HPLC)