MilliporeSigma
  • Common arterial trunk and ventricular non-compaction in Lrp2 knockout mice indicate a crucial role of LRP2 in cardiac development.
Common arterial trunk and ventricular non-compaction in Lrp2 knockout mice indicate a crucial role of LRP2 in cardiac development.
Disease models & mechanisms (2016-01-30)
Maria E Baardman, Mathijs V Zwier, Lambertus J Wisse, Adriana C Gittenberger-de Groot, Wilhelmina S Kerstjens-Frederikse, Robert M W Hofstra, Angelika Jurdzinski, Beerend P Hierck, Monique R M Jongbloed, Rolf M F Berger, Torsten Plösch, Marco C DeRuiter
ABSTRACT
Lipoprotein-related receptor protein 2 (LRP2) is important for development of the embryonic neural crest and brain in both mice and humans. Although a role in cardiovascular development can be expected, the hearts ofLrp2knockout (KO) mice have not yet been investigated. We studied the cardiovascular development ofLrp2KO mice between embryonic day 10.5 (E10.5) and E15.5, applying morphometry and immunohistochemistry, using antibodies against Tfap2α (neural crest cells), Nkx2.5 (second heart field), WT1 (epicardium derived cells), tropomyosin (myocardium) and LRP2. TheLrp2KO mice display a range of severe cardiovascular abnormalities, including aortic arch anomalies, common arterial trunk (persistent truncus arteriosus) with coronary artery anomalies, ventricular septal defects, overriding of the tricuspid valve and marked thinning of the ventricular myocardium. Both the neural crest cells and second heart field, which are essential for the lengthening and growth of the right ventricular outflow tract, are abnormally positioned in theLrp2KO. This explains the absence of the aorto-pulmonary septum, which leads to common arterial trunk and ventricular septal defects. Severe blebbing of the epicardial cells covering the ventricles is seen. Epithelial-mesenchymal transition does occur; however, there are fewer WT1-positive epicardium-derived cells in the ventricular wall as compared to normal, coinciding with the myocardial thinning and deep intertrabecular spaces. LRP2 plays a crucial role in cardiovascular development in mice. This corroborates findings of cardiac anomalies in humans withLRP2mutations. Future studies should reveal the underlying signaling mechanisms in which LRP2 is involved during cardiogenesis.
Sigma-Aldrich
Monoclonal Anti-Tropomyosin (Sarcomeric) antibody produced in mouse, clone CH1, ascites fluid