MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Study of genes associated with the 'anger-in' and 'anger-out' emotions of humans using a rat model.
Study of genes associated with the 'anger-in' and 'anger-out' emotions of humans using a rat model.
Experimental and therapeutic medicine (2015-03-18)
Yinghui Guo, Huiyun Zhang, Jie Gao, Sheng Wei, Chunhong Song, Peng Sun, Mingqi Qiao
ABSTRACT
The aim of the present study was to investigate the genes associated with 'anger-in' (tendency to suppress anger) and 'anger-out' (tendency to express anger through verbal or physical means) emotions in humans. Wistar rats were divided into five groups (n=10/group), based on the type of model and the Chinese medicinal formulation administered, and the rat models were established. The five groups were as follows: Normal control (control), anger-in model (AIM), anger-in Jingqianshu-administered (AIA), anger-out model (AOM) and anger-out Jingqianping-administered (AOA). Open-field, resident-intruder and aggressive behavior tests were carried out, as well as gene expression analysis, reverse transcription-quantitative polymerase chain reaction and western blot analyses. The body weights of the rats in the AIM and AOM groups were significantly lower than those of the control group rats. The open-field test indicated that the scores in the AOM group were significantly higher (P<0.05) than those in the AIM group. The aggression scores of the rats in the AOM group were significantly higher than those of the AIM group rats. Jingqianshu and Jingqianping granules attenuated the behavioral changes of the rats.
Sigma-Aldrich
Anti-GABAB Receptor 2 antibody produced in rabbit, affinity isolated antibody, lyophilized powder