MilliporeSigma
  • Tumor suppressor microRNA-34a inhibits cell proliferation by targeting Notch1 in renal cell carcinoma.
Tumor suppressor microRNA-34a inhibits cell proliferation by targeting Notch1 in renal cell carcinoma.
Oncology letters (2014-04-26)
Changcun Zhang, Ren Mo, Binde Yin, Libin Zhou, Yongchao Liu, Jie Fan
ABSTRACT
MicroRNA-34a (miR-34a) is a tumor suppressive microRNA, which induces G1 arrest, apoptosis and senescence by repressing the expression of multiple oncogenes. This study aimed to investigate the biological function and molecular mechanisms of miR-34a in human renal cell carcinoma (RCC) cells. Quantitative polymerase chain reaction (qPCR) revealed that miR-34a expression was significantly downregulated in eight of the 10 (80%) RCC tissues compared with adjacent normal tissues. In RCC cell lines, several other target genes of miR-34a were dysregulated at the mRNA level when the expression of miR-34a was elevated. In addition, western blot analysis and qPCR revealed that forced expression of miR-34a downregulated the expression of Notch1 at the protein and mRNA level. The Cell Counting Kit-8 identified that transient forced expression of miR-34a inhibited cell growth and resulted in cell cycle arrest, which was evaluated by flow cytometry. Our data demonstrated that miR-34a inhibits cell proliferation by downregulating Notch1 in RCC cell lines.