MilliporeSigma
  • Continuous green biocatalytic processes using ionic liquids and supercritical carbon dioxide.
Continuous green biocatalytic processes using ionic liquids and supercritical carbon dioxide.
Chemical communications (Cambridge, England) (2002-07-18)
Pedro Lozano, Teresa de Diego, Daniel Carrié, Michel Vaultier, José L Iborra
ABSTRACT
Soluble Candida antarctica lipase B dissolved in ionic liquids showed good synthetic activity, enantioselectivity and operational stability in supercritical carbon dioxide for both butyl butyrate synthesis and the kinetic resolution of 1-phenylethanol processes by transesterification.
Sigma-Aldrich
1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide, ≥97.0% (NMR)