MilliporeSigma
  • 3-Dimethylphosphinothioyl-2(3H)-oxazolone (MPTO), a promising new reagent for racemization-free couplings.
3-Dimethylphosphinothioyl-2(3H)-oxazolone (MPTO), a promising new reagent for racemization-free couplings.
International journal of peptide and protein research (1993-09-01)
T Katoh, M Ueki
ABSTRACT
3-Dimethylphosphinothioyl-2(3H)-oxazolone (MPTO) was synthesized, and its ability to effect racemization-free couplings and cyclization of a peptide and its C-terminal epimer was examined. MPTO showed good reactivity in aprotic polar solvents such as N,N-dimethylformamide (DMF) and N-methylpyrrolidone. In reactivity MPTO resembles DPPA and dimethylphosphinothioyl azide (MPTA) previously developed by us, but it is much better than these reagents because the side reactions specific to the azide method could be avoided. In coupling of Z-Gly-Val-OH with H-Val-OMe in DMF at 0 degree C, no racemization was observed without use of racemization-suppressing additives. Slight racemization observed at room temperature could be completely suppressed by addition of HOBt but not by HOSu. The utility of MPTO was demonstrated by the synthesis of cyclo-(D-Trp-D-Glu(OBzl)-Ala-D-Val-Leu), an intermediate for an endothelin-binding inhibitor BE 18257A. In a comparative study using DPPA, MPTA and MPTO, no racemization was observed for MPTA or MPTO, while DPPA caused considerable racemization. When MPTO was used in the presence of HOBt rapid cyclization (3 h at RT) occurred to give the optically pure cyclic product.
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, wetted with not less than 20 wt. % water, 97% dry basis
Pricing and availability is not currently available.
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, wetted with not less than 14 wt. % water, 98% dry basis
Pricing and availability is not currently available.