MilliporeSigma
  • Evaluation of intracellular lipids by standardized staining with a Sudan black B fraction.
Evaluation of intracellular lipids by standardized staining with a Sudan black B fraction.
Journal of biochemical and biophysical methods (1990-06-01)
H N Subramaniam, K A Chaubal
ABSTRACT
The selective staining of neutral lipids in Human Amnion cells in tissue culture was achieved using a particular fraction of the lipid dye, Sudan black B and a standardized staining procedure. The fraction, termed SBB-I, was isolated by thin-layer chromatography. The cytophotometric assessment of intracellular neutral lipids, stained with SBB-I, is described and applied to the study of changes in granulocytic neutral lipids in leukemia.
Sigma-Aldrich
Sudan Black B, certified by the BSC