MilliporeSigma
  • Do the long fatty acid chains of sphingolipids interdigitate across the center of a bilayer of shorter chain symmetric phospholipids?
Do the long fatty acid chains of sphingolipids interdigitate across the center of a bilayer of shorter chain symmetric phospholipids?
Biochimica et biophysica acta (1994-01-19)
J M Boggs, K M Koshy
ABSTRACT
Novel cerebroside sulfate (CBS) spin labels containing long chain C24 or C26 fatty acids with a nitroxide spin label on the 22nd carbon were synthesized and used to investigate the ability of the long fatty acid chains of glycosphingolipids to interdigitate across the center of a non-interdigitated bilayer of phospholipids formed of symmetric saturated or unsaturated shorter fatty acid chain species, in the presence or absence of cholesterol. The motion of these long chain spin labels incorporated at 1 mole% in dimyristoylphosphatidylcholine (diC14-PC), dipalmitoylphosphatidylcholine (diC16-PC), distearoylphosphatidylcholine (diC18-PC), dibehenoylphosphatidylcholine (diC22-PC), spingomyelin (SM), 1-stearoyl-2-oleoylphosphatidylcholine (18:0.18:1-PC), and dimyristoylphosphatidylethanolamine (diC14-PE) was compared to that of CBS spin labels containing stearic acid spin labeled at the 5th carbon and at the 16th carbon. The results indicated that the C26 chain is interdigitated in the gel phase of diC14-PC, diC16-PC, SM, and possibly diC18-PC, but not diC14-PE, and the C24 chain may interdigitate in diC14-PC but not in the other phospholipids. Thus in order to interdigitate across the center of gel phase bilayers, the long acyl chain of the sphingolipid probably must be long enough to nearly span the phospholipid bilayer. The inability to interdigitate in diC14-PE is likely due to the close packing of this lipid in the gel phase. The C26 chain may also be interdigitated in these lipids in the presence of cholesterol at low temperatures. However, at physiological temperatures in the presence of cholesterol and in the liquid-crystalline phase of all the lipids, the results indicate that the long acyl chain of the glycosphingolipid is not interdigitated, but rather must terminate at the bilayer center. This may force the carbohydrate headgroup of the glycosphingolipid farther above the bilayer surface, allowing it to be recognized better by various carbohydrate binding ligands and proteins.