MilliporeSigma
  • Scandium Trifluoromethanesulfonate as an Extremely Active Lewis Acid Catalyst in Acylation of Alcohols with Acid Anhydrides and Mixed Anhydrides.
Scandium Trifluoromethanesulfonate as an Extremely Active Lewis Acid Catalyst in Acylation of Alcohols with Acid Anhydrides and Mixed Anhydrides.
The Journal of organic chemistry (1996-07-12)
Kazuaki Ishihara, Manabu Kubota, Hideki Kurihara, Hisashi Yamamoto
ABSTRACT
Scandium trifluoromethanesulfonate (triflate), which is commercially available, is a practical and useful Lewis acid catalyst for acylation of alcohols with acid anhydrides or the esterification of alcohols by carboxylic acids in the presence of p-nitrobenzoic anhydrides. The remarkably high catalytic activity of scandium triflate can be used for assisting the acylation by acid anhydrides of not only primary alcohols but also sterically-hindered secondary or tertiary alcohols. The method presented is especially effective for selective macrolactonization of omega-hydroxy carboxylic acids.
Sigma-Aldrich
Scandium(III) triflate, 99%