MilliporeSigma
  • New indole derivatives as potent and selective serotonin uptake inhibitors.
New indole derivatives as potent and selective serotonin uptake inhibitors.
Journal of medicinal chemistry (1993-04-30)
J L Malleron, C Guérémy, S Mignani, J F Peyronel, A Truchon, J C Blanchard, A Doble, P Laduron, O Piot, J L Zundel
ABSTRACT
A series of new indole derivatives (2-28) has been prepared in the search for novel 5-HT uptake inhibitors. These compounds were obtained by the condensation of N-(chloroalkyl) naphthalenesultam derivatives with the appropriate amine in presence of a base, at reflux of DMF or THF. The yields were moderate (12-56%), except for the piperazine derivative 20 (85%). The affinity of the compounds for uptake site and 5-HT2, alpha 1, and D2 receptors was measured. Some compounds were studied in vivo by their potentiating effect of 5-HTP-induced symptomatology. The most potent and selective (uptake, 5-HT2 versus alpha 1, D2 sites) compounds contain a 3-[(4-piperidinyl)methyl]indole moiety. 5-Fluoro-3-[(4-piperidinyl)methyl]indole itself (compound 1) displayed a high affinity for the uptake site but was devoided of in vivo activity. N-Methylation of this compound abolished the affinity. In contrast N-substitution by a two-carbon chain linked to a naphthalenesultam or related heterocycle led to compounds exhibiting high affinity for the uptake site. One of them, 1-[2-[4-((5-fluoro-1H-indol-3-yl)methyl-1- piperidinyl]ethyl]-5,6-dihydro-1H,4H-1,2,5-thiadiazolo[4,3,2- ij]quinoline 2,2-dioxide (compound 24), was found as active as fluoxetine in vivo.
Sigma-Aldrich
4-Fluoroindole, 97%
Sigma-Aldrich
6-Fluoroindole, 98%