MilliporeSigma
Antibacterial activity of gentamicin sulfate in tissue culture.
Applied microbiology (1970-12-01)
A Rudin, A Healey, C A Phillips, D W Gump, B R Forsyth
ABSTRACT
Gentamicin is a more effective in vitro bacterial inhibitor than combined penicillinstreptomycin, is nontoxic to tissue culture monolayers, and does not inhibit virus replication.
Sigma-Aldrich
Gentamicin solution, 50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Gentamicin sulfate salt, powder, BioReagent, suitable for cell culture