MilliporeSigma
  • Rapid synthesis of nitrogen-doped graphene for a lithium ion battery anode with excellent rate performance and super-long cyclic stability.
Rapid synthesis of nitrogen-doped graphene for a lithium ion battery anode with excellent rate performance and super-long cyclic stability.
Physical chemistry chemical physics : PCCP (2013-11-30)
Tao Hu, Xiang Sun, Hongtao Sun, Guoqing Xin, Dali Shao, Changsheng Liu, Jie Lian
ABSTRACT
Chemical doping of nitrogen into graphene can significantly enhance the reversible capacity and cyclic stability of the graphene-based lithium ion battery (LIB) anodes, and first principles calculations based on density functional theory suggested that pyridinic-N shows stronger binding with Li with reduced energy barrier for Li diffusion and thus is more effective for Li storage than pyrrolic and graphitic-N. Here, we report a novel and rapid (~30 seconds) process to fabricate nitrogen-doped graphene (NGr) by simultaneous thermal reduction of graphene oxide with ammonium hydroxide. The porous NGr with dominant pyridinic N atoms displays greatly enhanced reversible capacities, rate performance and exceptional cyclic stability as compared with pristine graphene. The reversible discharge capacity of the NGr electrode cycled between 0.01-3 V can reach 453 mA h g(-1) after 550 cycles at a charge rate of 2 A g(-1) (~5.4 C), and 180 mA h g(-1) after 2000 cycles at a high charge rate of 10 A g(-1) (~27 C) without any capacity fading. When charged within 0.01-1.5 V, the NGr anode still exhibits high reversible capacities of 224 mA h g(-1) and 169 mA h g(-1) after 700 cycles and 800 cycles at a charge rate of 1 A g(-1) and 5 A g(-1), respectively. Ex situ X-ray photoelectron spectroscopy (XPS) analysis of the NGr electrode upon lithiation and delithiation indicated that the pyridinic-N dominates the capacity enhancement at 3 V, while the pyrrolic-N contributes primarily to Li ion storage below 1.5 V.