MilliporeSigma
  • G protein-coupled receptor kinase 5 gene polymorphisms are associated with postoperative atrial fibrillation after coronary artery bypass grafting in patients receiving β-blockers.
G protein-coupled receptor kinase 5 gene polymorphisms are associated with postoperative atrial fibrillation after coronary artery bypass grafting in patients receiving β-blockers.
Circulation. Cardiovascular genetics (2014-07-23)
Miklos D Kertai, Yen-Wei Li, Yi-Ju Li, Svati H Shah, William E Kraus, Manuel L Fontes, Mark Stafford-Smith, Mark F Newman, Mihai V Podgoreanu, Joseph P Mathew
ABSTRACT
We hypothesized that genetic variations in the adrenergic signaling pathway and cytochrome P450 2D6 enzyme are associated with new-onset atrial fibrillation (AF) in patients who underwent coronary artery bypass grafting and were treated with perioperative β-blockers (BBs). Two cohorts of patients who underwent coronary artery bypass grafting and received perioperative BBs at Duke University Medical Center were studied. In a discovery cohort of 563 individuals from the Perioperative Genetics and Safety Outcomes Study (PEGASUS), using a covariate-adjusted logistic regression analysis, we tested 492 single-nucleotide polymorphisms (SNPs) in 10 candidate genes of the adrenergic signaling pathway and cytochrome P450 2D6 for association with postoperative AF despite perioperative BB therapy. SNPs meeting a false discovery rate ≤0.20 (P<0.002) were then tested in the replication cohort of 245 individuals from the Catheterization Genetics biorepository. Of the 492 SNPs examined, 4 intronic SNPs of the G protein-coupled kinase 5 (GRK5) gene were significantly associated with postoperative AF despite perioperative BB therapy in the discovery cohort with additive odds ratios between 1.72 and 2.75 (P=4.78×10(-5) to 0.0015). Three of these SNPs met nominal significance levels in the replication cohort with odds ratios between 2.07 and 2.60 (P=0.007 to 0.016). However, meta-analysis of the 2 data sets cohorts suggested strong association with postoperative AF despite perioperative BB therapy in all 4 SNPs (meta-P values from 1.66×10(-6) to 3.39×10(-5)). In patients undergoing coronary artery bypass grafting, genetic variation in GRK5 is associated with postoperative AF despite perioperative BB therapy.
Sigma-Aldrich
GRK5, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution