MilliporeSigma
  • Loss of OMA1 delays neurodegeneration by preventing stress-induced OPA1 processing in mitochondria.
Loss of OMA1 delays neurodegeneration by preventing stress-induced OPA1 processing in mitochondria.
The Journal of cell biology (2016-01-20)
Anne Korwitz, Carsten Merkwirth, Ricarda Richter-Dennerlein, Simon E Tröder, Hans-Georg Sprenger, Pedro M Quirós, Carlos López-Otín, Elena I Rugarli, Thomas Langer
ABSTRACT
Proteolytic cleavage of the dynamin-like guanosine triphosphatase OPA1 in mitochondria is emerging as a central regulatory hub that determines mitochondrial morphology under stress and in disease. Stress-induced OPA1 processing by OMA1 triggersmitochondrial fragmentation, which is associated with mitophagy and apoptosis in vitro. Here, we identify OMA1 as a critical regulator of neuronal survival in vivo and demonstrate that stress-induced OPA1 processing by OMA1 promotes neuronal death and neuroinflammatory responses. Using mice lacking prohibitin membrane scaffolds as a model of neurodegeneration, we demonstrate that additional ablation of Oma1 delays neuronal loss and prolongs lifespan. This is accompanied by the accumulation of fusion-active, long OPA1 forms, which stabilize the mitochondrial genome but do not preserve mitochondrial cristae or respiratory chain supercomplex assembly in prohibitin-depleted neurons. Thus, long OPA1 forms can promote neuronal survival independently of cristae shape, whereas stress-induced OMA1 activation and OPA1 cleavage limit mitochondrial fusion and promote neuronal death.
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Spectrin alpha chain (nonerythroid) Antibody, clone AA6, clone AA6, Chemicon®, from mouse