MilliporeSigma
  • 5-Hydroxytryptamine promotes hepatocellular carcinoma proliferation by influencing β-catenin.
5-Hydroxytryptamine promotes hepatocellular carcinoma proliferation by influencing β-catenin.
Molecular oncology (2015-10-18)
Sarwat Fatima, Xiaoke Shi, Zesi Lin, Guo-qing Chen, Xiao-hua Pan, Justin Che-Yuen Wu, John W Ho, Nikki P Lee, Hengjun Gao, Ge Zhang, Aiping Lu, Zhao Xiang Bian
ABSTRACT
5-Hydroxytryptamine (5-HT), a neurotransmitter and vasoactive factor, has been reported to promote proliferation of serum-deprived hepatocellular carcinoma (HCC) cells but the detailed intracellular mechanism is unknown. As Wnt/β-catenin signalling is highly dysregulated in a majority of HCC, this study explored the regulation of Wnt/β-catenin signalling by 5-HT. The expression of various 5-HT receptors was studied by quantitative real-time polymerase chain reaction (qPCR) in HCC cell lines as well as in 33 pairs of HCC tumours and corresponding adjacent non-tumour tissues. Receptors 5-HT1D (21/33, 63.6%), 5-HT2B (12/33, 36.4%) and 5-HT7 (15/33, 45.4%) were overexpressed whereas receptors 5-HT2A (17/33, 51.5%) and 5-HT5 (30/33, 90.1%) were reduced in HCC tumour tissues. In vitro data suggests 5-HT increased total β-catenin, active β-catenin and decreased phosphorylated β-catenin protein levels in serum deprived HuH-7 and HepG2 cells compared to control cells under serum free medium without 5-HT. Activation of Wnt/β-catenin signalling was evidenced by increased expression of β-catenin downstream target genes, Axin2, cyclin D1, dickoppf-1 (DKK1) and glutamine synthetase (GS) by qPCR in serum-deprived HCC cell lines treated with 5-HT. Additionally, biochemical analysis revealed 5-HT disrupted Axin1/β-catenin interaction, a critical step in β-catenin phosphorylation. Increased Wnt/β-catenin activity was attenuated by antagonist of receptor 5-HT7 (SB-258719) in HCC cell lines and patient-derived primary tumour tissues in the presence of 5-HT. SB-258719 also reduced tumour growth in vivo. This study provides evidence of Wnt/β-catenin signalling activation by 5-HT and may represent a potential therapeutic target for hepatocarcinogenesis.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Supelco
Serotonin, analytical standard
Sigma-Aldrich
SB-258719, ≥98% (HPLC)