MilliporeSigma
  • Heparan sulfate 6-O-endosulfatases, Sulf1 and Sulf2, regulate glomerular integrity by modulating growth factor signaling.
Heparan sulfate 6-O-endosulfatases, Sulf1 and Sulf2, regulate glomerular integrity by modulating growth factor signaling.
American journal of physiology. Renal physiology (2016-01-15)
Yasutoshi Takashima, Kazuko Keino-Masu, Hiroshi Yashiro, Satoshi Hara, Tomo Suzuki, Toin H van Kuppevelt, Masayuki Masu, Michio Nagata
ABSTRACT
Glomerular integrity and functions are maintained by growth factor signaling. Heparan sulfate, the major component of glomerular extracellular matrixes, modulates growth factor signaling, but its roles in glomerular homeostasis are unknown. We investigated the roles of heparan sulfate 6-O-endosulfatases, sulfatase (Sulf)1 and Sulf2, in glomerular homeostasis. Both Sulf1 and Sulf2 were expressed in the glomeruli of wild-type (WT) mice. Sulf1 and Sulf2 double-knockout (DKO) mice showed glomerular hypercellularity, matrix accumulation, mesangiolysis, and glomerular basement membrane irregularity. Platelet-derived growth factor (PDGF)-B and PDGF receptor-β were upregulated in Sulf1 and Sulf2 DKO mice compared with WT mice. Glomeruli from Sulf1 and Sulf2 DKO mice in vitro stimulated by either PDGF-B, VEGF, or transforming growth factor-β similarly showed reduction of phospho-Akt, phospho-Erk1/2, and phospho-Smad2/3, respectively. Since glomerular lesions in Sulf1 and Sulf2 DKO mice were reminiscent of diabetic nephropathy, we examined the effects of Sulf1 and Sulf2 gene disruption in streptozotocin-induced diabetes. Diabetic WT mice showed an upregulation of glomerular Sulf1 and Sulf2 mRNA by in situ hybridization. Diabetic DKO mice showed significant increases in albuminuria and serum creatinine and an acceleration of glomerular pathology without glomerular hypertrophy; those were associated with a reduction of glomerular phospho-Akt. In conclusion, Sulf1 and Sulf2 play indispensable roles to maintain glomerular integrity and protective roles in diabetic nephropathy, probably by growth factor modulation.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Platelet-Derived Growth Factor-BB human, PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture