MilliporeSigma
  • Cyclic RGD peptidomimetics containing 4- and 5-amino-cyclopropane pipecolic acid (CPA) templates as dual αVβ3 and α5β1 integrin ligands.
Cyclic RGD peptidomimetics containing 4- and 5-amino-cyclopropane pipecolic acid (CPA) templates as dual αVβ3 and α5β1 integrin ligands.
Bioorganic & medicinal chemistry (2016-01-13)
Lorenzo Sernissi, Andrea Trabocchi, Dina Scarpi, Francesca Bianchini, Ernesto G Occhiato
ABSTRACT
4-Amino- and 5-amino-cyclopropane pipecolic acids (CPAs) with cis relative stereochemistry between the carboxylic and amino groups were used as templates to prepare cyclic peptidomimetics containing the RGD sequence as possible integrin binders. The peptidomimetic c(RGD8) built on the 5-amino-CPA displayed an inhibition activity (IC50=2.4nM) toward the αvβ3 integrin receptor (expressed in M21 human melanoma cell line) comparable to that of the most potent antagonists reported so far and it was ten times more active than the corresponding antagonist c(RGD7) derived from the isomeric 4-amino-CPA. Both compounds were also nanomolar ligands of the α5β1 integrin (expressed in human erythroleukemia cell line K562). These results suggest that the CPA-derived templates are suitable for the preparation of dual αvβ3 and α5β1 ligands to suppress integrin-mediated events as well as for targeted drug delivery in cancer therapy.
Sigma-Aldrich
Vitronectin from human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-(−)-Glucose, ≥99%