MilliporeSigma
An apolipoprotein B100 mimotope prevents obesity in mice.
Clinical science (London, England : 1979) (2015-11-01)
Hyo Joon Kim, Hee Jong Lee, Jung Soon Choi, Jemin Han, Ji Young Kim, Hyun Kyun Na, Hae-Jung Joung, Young Sik Kim, Bert Binas
ABSTRACT
Although apolipoprotein B100 (ApoB100) plays a key role in peripheral fat deposition, it is not considered a suitable therapeutic target in obesity. In the present study we describe a novel ApoB100 mimotope, peptide pB1, and the use of pB1-based vaccine-like formulations (BVFs) against high-fat diet (HFD)-induced obesity. In HFD- compared with chow-fed adolescent mice, BVFs reduced the 3-month body-weight gains attributable to increased dietary fat by 44-65%, and prevented mesenteric fat accumulation and liver steatosis. The body-weight reductions paralleled the titres of pB1-reactive immunoglobulin G (IgG) antibodies, and pB1-reactive antibodies specifically recognized native ApoB100 and a synthetic peptide from the C-terminal half of ApoB100. In cultured 3T3L1 adipocytes, anti-pB1 antibodies increased lipolysis and inhibited low-density lipoprotein (LDL) uptake. In cultured RAW 264.7 macrophages, the same antibodies enhanced LDL uptake (without causing foam cell formation). These findings make ApoB100 a promising target for an immunization strategy against HFD-induced obesity.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Apolipoprotein B, Human Plasma, Low-Density Lipoprotein, Native apolipoprotein B from human plasma. It is a dominant protein constituent of LDL and VLDL. Acts as a ligand for LDL receptor, directing clearance of LDL from plasma to the liver.