MilliporeSigma
  • Harnessing Enzymatic Promiscuity in Myxochelin Biosynthesis for the Production of 5-Lipoxygenase Inhibitors.
Harnessing Enzymatic Promiscuity in Myxochelin Biosynthesis for the Production of 5-Lipoxygenase Inhibitors.
Chembiochem : a European journal of chemical biology (2015-09-30)
Juliane Korp, Stefanie König, Sebastian Schieferdecker, Hans-Martin Dahse, Gabriele M König, Oliver Werz, Markus Nett
ABSTRACT
The siderophore myxochelin A is a potent inhibitor of human 5-lipoxygenase (5-LO). To clarify whether the iron-chelating properties of myxochelin A are responsible for this activity, several analogues of this compound were generated in the native producer Pyxidicoccus fallax by precursor-directed biosynthesis. Testing in a cell-free assay unveiled three derivatives with bioactivity comparable with that of myxochelin A. Furthermore, it became evident that inhibition of 5-LO by myxochelins does not correlate with their iron affinities.