MilliporeSigma
Organizing junctions at the cell-cell interface.
Cell (2008-12-02)
Anna Akhmanova, Alpha S Yap
ABSTRACT
The zonula adherens (ZA) is a specialized cadherin-based structure found at the contacts between epithelial cells. Meng et al. (2008) now identify a protein complex containing the microtubule minus-end-binding protein Nezha, which provides a critical link between microtubules and cadherins in ZA biogenesis and maintenance.