MilliporeSigma
A role for retromer in hepatitis C virus replication.
Cellular and molecular life sciences : CMLS (2015-08-25)
Peiqi Yin, Zhi Hong, Xiaojie Yang, Raymond T Chung, Leiliang Zhang
ABSTRACT
Hepatitis C virus (HCV) has infected over 170 million people worldwide. Phosphatidylinositol 4-phosphate (PI4P) is the organelle-specific phosphoinositide enriched at sites of HCV replication. Whether retromer, a PI4P-related host transport machinery, unloads its cargo at HCV replication sites remains inconclusive. We sought to characterize the role of retromer in HCV replication. Here, we demonstrated the interaction between retromer subunit Vps35 and HCV NS5A protein by immunoprecipitation and GST pulldown. Vps35 colocalized with NS5A and PI4P in both OR6 replicon and JFH1 infected Huh 7.5.1 cells. HCV replication was inhibited upon silencing retromer subunits. CIMPR, a typical retromer cargo, participated in HCV replication. Our data suggest that retromer component Vps35 is recruited by NS5A to viral replication sites where PI4P unloads CIMPR. These findings demonstrate a dependence role of retromer in HCV replication and identify retromer as a potential therapeutic target against HCV.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
MISSION® esiRNA, targeting human VPS35