MilliporeSigma
  • Non-Hematopoietic β-Arrestin1 Confers Protection Against Experimental Colitis.
Non-Hematopoietic β-Arrestin1 Confers Protection Against Experimental Colitis.
Journal of cellular physiology (2015-10-20)
Taehyung Lee, Eunhee Lee, David Arrollo, Peter C Lucas, Narayanan Parameswaran
ABSTRACT
β-Arrestins are multifunctional scaffolding proteins that modulate G protein-coupled receptor (GPCR)-dependent and -independent cell signaling pathways in various types of cells. We recently demonstrated that β-arrestin1 (β-arr1) deficiency strikingly attenuates dextran sodium sulfate (DSS)-induced colitis in mice. Since DSS-induced colitis is in part dependent on gut epithelial injury, we examined the role of β-arr1 in intestinal epithelial cells (IECs) using a colon epithelial cell line, SW480 cells. Surprisingly, we found that knockdown of β-arr1 in SW480 cells enhanced epithelial cell death via a caspase-3-dependent process. To understand the in vivo relevance and potential cell type-specific role of β-arr1 in colitis development, we generated bone marrow chimeras with β-arr1 deficiency in either the hematopoietic or non-hematopoietic compartment. Reconstituted chimeric mice were then subjected to DSS-induced colitis. Similar to our previous findings, β-arr1 deficiency in the hematopoietic compartment protected mice from DSS-induced colitis. However, consistent with the role of β-arr1 in epithelial apoptosis in vitro, non-hematopoietic β-arr1 deficiency led to an exacerbated colitis phenotype. To further understand signaling mechanisms, we examined the effect of β-arr1 on TNF-α-mediated NFκB and MAPK pathways. Our results demonstrate that β-arr1 has a critical role in modulating ERK, JNK and p38 MAPK pathways mediated by TNF-α in IECs. Together, our results show that β-arr1-dependent signaling in hematopoietic and non-hematopoietic cells differentially regulates colitis pathogenesis and further demonstrates that β-arr1 in epithelial cells inhibits TNF-α-induced cell death pathways.