MilliporeSigma
  • Chronic acidosis in the tumour microenvironment selects for overexpression of LAMP2 in the plasma membrane.
Chronic acidosis in the tumour microenvironment selects for overexpression of LAMP2 in the plasma membrane.
Nature communications (2015-12-15)
Mehdi Damaghi, Narges K Tafreshi, Mark C Lloyd, Robert Sprung, Veronica Estrella, Jonathan W Wojtkowiak, David L Morse, John M Koomen, Marilyn M Bui, Robert A Gatenby, Robert J Gillies
ABSTRACT
Early cancers are avascular and hence, profoundly acidic. Pre-malignant cells must adapt to acidosis to thrive in this hostile microenvironment. Here, we investigate MCF-7 cells that are adapted to grow in acidic conditions using SILAC proteomics and we reveal a significant upregulation of lysosomal proteins. Prominent among these is LAMP2 that functions to protect lysosomal membranes from acid proteolysis. LAMP2 upregulation by acidosis is confirmed both in vitro and in vivo. Furthermore, we show that the depletion of LAMP2 is sufficient to increase acidosis-mediated toxicity. In breast cancer patient samples, there is a high correlation of LAMP2 mRNA and protein expression with progression. We also observe that LAMP2 is located at the plasma membrane in clinical samples and this redistribution is acid-induced in vitro. Our findings suggest a potential adaptive mechanism, wherein cells chronically exposed to an acidic environment translocate lysosomal proteins to their surface, thus protecting the plasmalemma from acid-induced hydrolysis.
Sigma-Aldrich
Allyl methyl sulfone, 96%
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-S100A6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution