MilliporeSigma
  • Rapid Software-Based Design and Optical Transient Liquid Molding of Microparticles.
Rapid Software-Based Design and Optical Transient Liquid Molding of Microparticles.
Advanced materials (Deerfield Beach, Fla.) (2015-10-29)
Chueh-Yu Wu, Keegan Owsley, Dino Di Carlo
ABSTRACT
Microparticles with complex 3D shape and composition are produced using a novel fabrication method, optical transient liquid molding, in which a 2D light pattern exposes a photopolymer precursor stream shaped along the flow axis by software-aided inertial flow engineering.
Sigma-Aldrich
Poly(ethylene glycol) diacrylate, average Mn 575
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Hydroxy-2-methylpropiophenone, 97%